Learn More
Trimethylamine hydrochloride, 98%, Thermo Scientific Chemicals
13.36€ - 35.66€
Chemical Identifiers
CAS | 593-81-7 |
---|---|
Molecular Formula | C3H10ClN |
Molecular Weight (g/mol) | 95.57 |
MDL Number | MFCD00012478 |
InChI Key | SZYJELPVAFJOGJ-UHFFFAOYSA-N |
Synonym | trimethylamine hydrochloride, trimethylammonium chloride, trimethylamine hcl, n,n-dimethylmethanamine hydrochloride, trimethylamine monohydrochloride, methanamine, n,n-dimethyl-, hydrochloride, trimethylaminehydrochloride, unii-uw38srm77k, uw38srm77k, trimethyl ammonium chloride |
PubChem CID | 10313079 |
ChEBI | CHEBI:64700 |
IUPAC Name | N,N-dimethylmethanamine;hydrochloride |
SMILES | [H+].[Cl-].CN(C)C |
Description
Trimethylamine hydrochloride is used in the manufacture of quaternary ammonium compounds, as insect attractant. It is used as a raw material for the synthesis of choline chloride, quaternary ammonium compounds, ion exchange resins. It acts as cationic starch reagents, phase transfer catalyst, oil field chemicals, warning agent for natural gas and flotation agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTrimethylamine hydrochloride is used in the manufacture of quaternary ammonium compounds, as insect attractant. It is used as a raw material for the synthesis of choline chloride, quaternary ammonium compounds, ion exchange resins. It acts as cationic starch reagents, phase transfer catalyst, oil field chemicals, warning agent for natural gas and flotation agent.
Solubility
Soluble in water (737 mg/ml at 20°C), chloroform (moderately), and alcohol. Insoluble in ether.
Notes
Hygroscopic. Store under dry inert gas and away from moisture. Incompatible with oxidizing agents and water.
Chemical Identifiers
593-81-7 | |
95.57 | |
SZYJELPVAFJOGJ-UHFFFAOYSA-N | |
10313079 | |
N,N-dimethylmethanamine;hydrochloride |
C3H10ClN | |
MFCD00012478 | |
trimethylamine hydrochloride, trimethylammonium chloride, trimethylamine hcl, n,n-dimethylmethanamine hydrochloride, trimethylamine monohydrochloride, methanamine, n,n-dimethyl-, hydrochloride, trimethylaminehydrochloride, unii-uw38srm77k, uw38srm77k, trimethyl ammonium chloride | |
CHEBI:64700 | |
[H+].[Cl-].CN(C)C |
Specifications
593-81-7 | |
C3H10ClN | |
3905063 | |
14,9710 | |
Soluble in water (737mg/ml at 20°C),chloroform (moderately),and alcohol. Insoluble in ether. | |
[H+].[Cl-].CN(C)C | |
95.57 | |
CHEBI:64700 | |
98% |
∽280°C (decomposition) | |
MFCD00012478 | |
Hygroscopic | |
trimethylamine hydrochloride, trimethylammonium chloride, trimethylamine hcl, n,n-dimethylmethanamine hydrochloride, trimethylamine monohydrochloride, methanamine, n,n-dimethyl-, hydrochloride, trimethylaminehydrochloride, unii-uw38srm77k, uw38srm77k, trimethyl ammonium chloride | |
SZYJELPVAFJOGJ-UHFFFAOYSA-N | |
N,N-dimethylmethanamine;hydrochloride | |
10313079 | |
95.57 | |
Trimethylamine hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 209-810-0
missing translation for 'rtecsNumber' : YH2700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only