missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Trimethoprim, 98%

Product Code. 15780889
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Trimethoprim, CAS # 738-70-5, is a trimethoxybenzyl-pyrimidine used as an antimicrobial.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Trimethoprim is a synthetic pyrimidine analog that binds dihydrofolate-reductase and inhibits the synthesis of folic acid; when paired with sulfamethoxazole, its activity is enhanced • Trimethoprim has antibacterial and antiprotozoal properties

Applications

• Trimethoprim inhibits the conversion of para-aminobenzoic acid (PABA) to folic acid near the end of the synthesis pathway and can be used to study related effects

• Trimethoprim has been radiolabeled and used to trace routes of infection, and it also has been used experimentally as a fluorescent labeler to quantify dihydrofolate-reductase expression

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 738-70-5
Molecular Formula C14H18N4O3
Molecular Weight (g/mol) 290.32
InChI Key IEDVJHCEMCRBQM-UHFFFAOYSA-N
Synonym trimethoprim, proloprim, trimpex, bactramin, monotrim, monotrimin, trimetoprim, trimopan, monoprim, syraprim
PubChem CID 5578
ChEBI CHEBI:45924
IUPAC Name 5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
SMILES COC1=CC(=CC(=C1OC)OC)CC2=CN=C(N=C2N)N

Specifications

Melting Point 234.0°C to 236.0°C
Loss on Drying 0.5% max. (105°C, 4 h)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Solubility Information Solubility in water: insoluble.
Formula Weight 290.32
Percent Purity 98%
Chemical Name or Material Trimethoprim
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.