missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylene Glycol Dimethyl Ether, 99%, Reagent Grade, Honeywell™
Click to view available options
Quantity:
250 g
1 L
Packaging:
Glass bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Melting Point | -45°C |
Density | 0.986g/mL at 25°C |
Boiling Point | 216°C |
Flash Point | 113°C (235.4°F) |
Ignition Point | 375°F |
Packaging | Glass bottle |
Linear Formula | CH3O(CH2CH2O)3CH3 |
Quantity | 1 L |
UN Number | NONH for all modes of transport |
Beilstein | 1700630 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'