missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Fluka™ Triethylammonium Formate Solution, Honeywell™
volatile buffer, 1 M pH 6.0, for HPLC
76.87€ - 342.70€
Specifications
Color | Colorless |
---|---|
For Use With (Equipment) | Volatile buffer for HPLC |
Molecular Weight (g/mol) | 147.218 |
pH | 5.9 to 6.1 (at 25°C) |
Packaging | Plastic Bottle |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Colorless | |
147.218 | |
Plastic Bottle | |
Liquid |
Volatile buffer for HPLC | |
5.9 to 6.1 (at 25°C) | |
585-29-5 |