Learn More
Copan Italia™ Cell Spreader
Use the Copan Italia™ Cell Spreader to perform bacterial CFU plate counts on water, milk, urine and other liquid samples.The laboratory apparatus is disposable, sterile, T-shaped and blue in color.
Brand:  Copan Italia™ 173CS10
 View more versions of this product
Product Code. 11716638
  • 183.00€ / Pack of 1000
Estimated Shipment: 30-12-2024
to see stock.
Explore more special offers
Description
  • 171mm
  • T shape
  • Sterile
  • Blue
Specifications
Specifications
Single-use
171 mm
T
Cell Spreader
Blue
1000/Pk.
Polystyrene