missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butylphosphine, 95%, Thermo Scientific Chemicals

Product Code. 10306740
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 998-40-3
Molecular Formula C12H27P
Molecular Weight (g/mol) 202.32
MDL Number MFCD00009462
InChI Key TUQOTMZNTHZOKS-UHFFFAOYSA-N
Synonym tributylphosphine, tri-n-butylphosphine, phosphine, tributyl, tris butyl phosphine, tributylfosfin, tributylfosfin czech, tributyl-phosphane, unii-0o52fjr7wn, tri-n-butyl phosphine, tributyl phosphine tbp
PubChem CID 13831
IUPAC Name tributylphosphane
SMILES CCCCP(CCCC)CCCC

Specifications

Melting Point -65.0°C
Color Colorless to Yellow
Density 0.8100g/mL
Boiling Point 240.0°C to 242.0°C
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula [CH3(CH2)3]3P
Refractive Index 1.4610 to 1.4630
Quantity 500 mL
Beilstein 04, II, 971
Fieser 01,1191; 05,685; 15,325; 17,269; 06,604; 07,738; 13,158
Specific Gravity 0.81
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with ether,methanol,ethanol and benzene
Formula Weight 202.32
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Tri-n-butylphosphine, 95%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tri-n-butylphosphine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Pyrophoric liquid Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H250-Catches fire spontaneously if exposed to air.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.