missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TREM1 Antibody (174031), Allophycocyanin/Cy7, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1187APCCY7
This item is not returnable.
View return policy
Description
TREM1 Monoclonal antibody specifically detects TREM1 in Mouse samples. It is validated for Flow Cytometry
Specifications
| TREM1 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 174031 | |
| Flow Cytometry | |
| CD354 antigen, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TREM-1, Ala21-Ser202, Accession # Q9JKE2 | |
| 0.1 mL | |
| Signal Transduction | |
| 54210 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction