Learn More
Deutsch Neumann™ Transparent Polyethylene Tubings
Click to view available options
Diameter (Metric) Inner:
4 mm
Unit Size:
100m
Specifications
Specifications
| Diameter (Metric) Inner | 4 mm |
| Material | Polyethylene |
| Thickness (Metric) | 1 mm |
| Color | Transparent |