Learn More
VITLAB™ Narrow Mouth LDPE Safety Wash Bottles
Product Code. 11867900
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Chemical Label:
Acetic Acid
Acetonitrile
Distilled Water
Ethanol
Ethyl Acetate
Heptane
Isopropanol
Methanol
N,N-Dimethylformamide
Pentane
Tetrahydrofuran (THF)
Toluene
Xylene
Screw Color:
Blue
Green
Orange
Red
Yellow
Thread Size (Metric):
GL 25
GL 32
Unit Size:
Pack of 12
Pack of 6
Product Code. 11867900
Brand: VITLAB™ 1332839
Only null left
Add to basket
Add to basket
Work safely with potentially dangerous chemicals. VITLAB™ VITsafe™ Narrow Mouth LDPE Safety Wash Bottles feature a permanent safety imprint, color-coded VENT-CAP and precise spray control to eliminate drips and leaks and ensure safe handling.
VITsafe™ Narrow Mouth LDPE Safety Wash Bottles – Wash bottles meet all safety requirements for working with potentially dangerous chemicals.
Specifications
Capacity (Metric) 500 mL
Color Transparent
Material LDPE, Polypropylene
Packaging Individually wrapped in PE bag
Quantity 12Pack
Screw Color Green
Thread Size (Metric) GL 25
Mouth Narrow
Height (Metric) 180 mm
Product Type Safety Wash Bottle
Chemical Label Methanol
Diameter (Metric) 74 mm
Closure Type Vented Screw Cap