missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ TranslationBlocker Human and Mouse Apoptosis regulator Bcl-2 siRNA

Product Code. 16069161
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
This item is not returnable. View return policy

Product Code. 16069161

Brand: enQuireBio™ QX1010nmol

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Blocks transcription and translation of Apoptosis regulator Bcl-2 from Human and Mouse.

Specifications

Accession Number NP_000648
Cell Lines Tested Hct116 Colorectal Carcinoma Cells.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol BCL2
Gene ID (Entrez) 596
Product Type siRNA
Quantity 10 nmol
Synonym Bcl-2, PPP1R50
Target Human, Mouse
Form Lyophilized
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.