missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-Ned 19, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
Nicotinic acid adenine dinucleotide phosphate (NAADP) antagonist. Inhibits NAADP-mediated Ca2+ release (IC50 = 6 nM); also inhibits [32P]NAADP binding (IC50 = 0.4 nM). Fluorescently labels NAADP receptors in intact cells. Stereosiomer of cis-Ned 19 (Cat. No. 3953).
Specifications
Specifications
CAS | 1354235-96-3 |
Molecular Formula | C30H31FN4O3 |
Quantity | 10 mg |
Synonym | trans-ned 19, ned-19, trans-ned-19, 1r,3s-1-3-4-2-fluorophenyl piperazin-1-yl methyl-4-methoxyphenyl-2,3,4,9-tetrahydro-1h-pyrido 3,4-b indole-3-carboxylic acid, naadp antagonist, nicotinic acid adenine dinucleotide phosphate antagonist, 1r,3s-1-3-4-2-fluorophenyl piperazin-1-yl methyl-4-methoxyphenyl-2,3,4,9-tetrahydro-1h-beta-carboline-3-carboxylic acid |
InChI Key | FUHCEERDBRGPQZ-LBNVMWSVSA-N |
SMILES | COC1=C(C=C(C=C1)C2C3=C(CC(N2)C(=O)O)C4=CC=CC=C4N3)CN5CCN(CC5)C6=CC=CC=C6F |
IUPAC Name | (1R,3S)-1-[3-[[4-(2-fluorophenyl)piperazin-1-yl]methyl]-4-methoxyphenyl]-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylic acid |
Molecular Weight (g/mol) | 514.601 |
PubChem CID | 1427628 |
Formula Weight | 514.59 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'