missing translation for 'onlineSavingsMsg'
Learn More

trans-N-Boc-4-cyano-L-proline methyl ester, 97%, Thermo Scientific Chemicals

Product Code. 15489467
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 194163-91-2
Molecular Formula C12H18N2O4
Molecular Weight (g/mol) 254.29
MDL Number MFCD03094915
InChI Key GHLKJIQVORAVHE-UHFFFAOYNA-N
Synonym boc-trans-4-cyano-l-proline methyl ester, 2s,4r-1-tert-butyl 2-methyl 4-cyanopyrrolidine-1,2-dicarboxylate, n-boc-trans-4-cyano-l-proline methyl ester, 1-tert-butyl 2-methyl 2s,4r-4-cyanopyrrolidine-1,2-dicarboxylate, n-tert-butoxycarbonyl-trans-4-cyano-l-proline methyl ester, 4r-1-tert-butyloxycarbonyl-4-cyano-l-proline methyl ester, 2s,4r-1-tert-butoxycarbonyl-4-cyano-2-methylpyrrolidine-2-carboxylic acid, 1,2-pyrrolidinedicarboxylicacid,4-cyano-,1-1,1-dimethylethyl 2-methyl ester, 2s,4r, 1,2-pyrrolidinedicarboxylic acid, 4-cyano-, 1-1,1-dimethylethyl 2-methyl ester, 2s,4r, 1,2-pyrrolidinedicarboxylicacid, 4-cyano-, 1-1,1-dimethylethyl 2-methyl ester, 2s,4r
PubChem CID 17998966
IUPAC Name 1-O-tert-butyl 2-O-methyl (2S,4R)-4-cyanopyrrolidine-1,2-dicarboxylate
SMILES COC(=O)C1CC(CN1C(=O)OC(C)(C)C)C#N

Specifications

Quantity 1 g
Formula Weight 254.29
Percent Purity 97%
Chemical Name or Material trans-N-Boc-4-cyano-L-proline methyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'