missing translation for 'onlineSavingsMsg'
Learn More

(±)-trans-ACPD, Tocris Bioscience™

Product Code. 30001057
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Group I and II mGlu agonist

Equimolecular mixture of (1S,3R)- and (1R,3S)-ACPD. Selective agonist for metabotropic glutamate receptors; active at both group I and group II mGlu receptors (EC50 values are 2, 15, 23 and ∽800 μM at mGluR2, mGluR1, mGluR5 and mGluR4 respectively).

(1S,3R)-ACPD and cis-ACPD also available.

Chemical Identifiers

CAS 67684-64-4
Molecular Formula C7H11NO4
Molecular Weight (g/mol) 173.168
InChI Key YFYNOWXBIBKGHB-FFWSUHOLSA-N
Synonym 1s,3s-1-aminocyclopentane-1,3-dicarboxylic acid, chembl29726, trans-acpd, cis-acpd, +/--trans-acpd, 1s, 3r-acpd, 1-amino-cis-cyclopentane-1,3-dicarboxylic acid, tocris-0186, tocris-0187, tocris-0284
PubChem CID 6604704
IUPAC Name (1S,3S)-1-aminocyclopentane-1,3-dicarboxylic acid
SMILES C1CC(CC1C(=O)O)(C(=O)O)N

Specifications

Quantity 10 mg
Formula Weight 173.17
Percent Purity >99%
Chemical Name or Material (±)-trans-ACPD
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'