missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-trans-ACPD, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Equimolecular mixture of (1S,3R)- and (1R,3S)-ACPD. Selective agonist for metabotropic glutamate receptors; active at both group I and group II mGlu receptors (EC50 values are 2, 15, 23 and ∽800 μM at mGluR2, mGluR1, mGluR5 and mGluR4 respectively).
(1S,3R)-ACPD and cis-ACPD also available.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 173.17 |
Percent Purity | >99% |
Chemical Name or Material | (±)-trans-ACPD |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'