missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tranilast, ACROS Organics™

66.90€ - 206.00€

Specifications

Chemical Name or Material Tranilast
CAS 53902-12-8
Solubility insoluble in water
Merck Index 9646
Melting Point 166.2°C to 168.2°C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17392481
View Documents
Thermo Scientific™
468610010
1 g
66.90€
1g
Please to purchase this item. Need a web account? Register with us today!
17302491
View Documents
Thermo Scientific™
468610050
5 g
206.00€
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Tranilast
insoluble in water
166.2°C to 168.2°C
327.33
White to Light-yellow to Green
53902-12-8
9646
C18 H17 N O5
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS07: Exclamation mark

Recommended Storage : Refrigerator +4°C

SDS
Documents

Documents

Product Certifications