missing translation for 'onlineSavingsMsg'
Learn More

TPL 10 Trace Model Buffer Solution pH 10.1, WTW™

Product Code. 10643313
Click to view available options
Quantity:
250 mL
Packaging:
Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

pH buffer by WTW™ that is precise and traceable to PTB/NIST in a bulk size of 250 ml with an accuracy that is ± 0,03 pH.

  • pH buffer by WTW that is precise and traceable to PTB/NIST in a bulk size of 250 ml 
  • Live up to 3 years (unopened)
  • Thrifty use
  • Easiest handling by means of dosing device
  • Buffer 10.0 is only still suitable for inoLab™ device of the series level 1, 2, 3, portable meters 3xx as well as for various other old devices!
  • Modern technical buffering solution is traceable to NIST (National Institute of Standards and Technology, USA) 
  • Accuracy is ± 0,03 pH
TRUSTED_SUSTAINABILITY

Specifications

pH 10.1
Physical Form Liquid
Quantity 250 mL
Packaging Bottle
Traceability to NIST Yes
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'