Learn More
Invitrogen™ ToxBLAzer™ DualScreen Kit
ToxBLAzer™ DualScreen Kit
1530.00€ - 32860.00€
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10624223
View Documents
Invitrogen™
K1138
200 μg
1530.00€
Each
In Stock
to see stock.
Add to basket
11504936
View Documents
Invitrogen™
K1139
5mg
32860.00€
5mg
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
Description
Description
ToxBLAzer™ DualScreen Kit provide the key advantage of combining the ratiometric reporter gene readout of beta-lactamase with a cytotoxicity readout in the same assay. Thus, ToxBLAzer™ DualScreen yields reporter readouts that normalize cytotoxic cellular responses, which is a necessity in screening applications which down regulate transcription. The ToxBLAzer™ substrate contains both the LiveBLAzer™-FRET B/G substrate and a proprietary cytotoxicity indicator. Both compounds readily enter the cell, where endogenous esterases rapidly convert them into fluorescent compounds Esterase activity and an intact cell membrane are required for detection of the indicator. The cytotoxicity indicator's fluorescence is red-shifted to ensure compatibility with the blue and green emissions of the LiveBLAzer™-FRET B/G substrate, allowing detection of both reporter gene activity and cytotoxicity in the same assay sample.
Advantages of ToxBLAzer™ DualScreen Kits:
  • Multiplexed readout results in rapid identification of false positives due to toxicity or lack of viable cells, thereby improving hit-picking efficiency
  • Ratiometric blue:green reporter readout reduces false hits by minimizing experimental noise
  • Single reagent addition eliminates need for follow up cytotoxicity assays, thereby increasing throughput while reducing sample and test compound consumption
  • Videos