missing translation for 'onlineSavingsMsg'
Learn More

SCA Hygiene Products™ Tork Fibre Basic Paper

Brand:  SCA Hygiene Products™ 121206_CLEAN

 View more versions of this product

Product Code. 10718425

  • 96.00€ / Pack of 6
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Basic Paper
200 mm
ISO 9001, ISO 14001 and EMAS
Single-use
Centerfeed Roll
6 Roll
160 m
Fiber
White
160 m x 200 mm
Tork Centre feed Dispenser, Tork Centre feed Starter Pack, Tork Metal Centre feed Dispenser
2-ply
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions