Learn More
Emma Safety Footwear Torino Safety Shoes
The formally styled business shoe that protects the feet from above and below with water-resistant leather uppers, zero penetration in-sole and a steel safety toe cap. Specifically designed for standing, walking and the occapional run.
184.00€
Specifications
Certifications/Compliance | EN ISO 20345:2011, EN 12568:2010 |
---|---|
Closure Type | Laces |
Color | Black |
Design | Torino |
Liner Material | Hydro-Tec™ Sanitized Silver & Wellness leather |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
13204454
|
Emma Safety Footwear
MM13509040 |
40 |
184.00€
1 pair |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
13214454
|
Emma Safety Footwear
MM13509041 |
41 |
184.00€
1 pair |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
13264454
|
Emma Safety Footwear
MM13509046 |
46 |
184.00€
1 pair |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
13284454
|
Emma Safety Footwear
MM13509048 |
48 |
184.00€
1 pair |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction
- Nitrile Rubber Heat Resistant Outsole
- ESD Certified
- Chrome-free lining
- Sanitized
Specifications
EN ISO 20345:2011, EN 12568:2010 | |
Black | |
Hydro-Tec™ Sanitized Silver & Wellness leather | |
Steel |
Laces | |
Torino | |
Leather |