missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Tolvaptan
GREENER_CHOICE

Product Code. p-5546079
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18724848

Brand: Tocris Bioscience™ 5181/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective vasopressin V2 receptor antagonist; renoprotective and orally active

Tolvaptan is a potent and selective competitive vasopressin V2 receptor antagonist (Ki values are 0.06 and 12.3 nM for V2 and V1a receptors respectively). Decreases urine osmolality and increases serum sodium concentrations. Delays the onset of end-stage renal disease in a mouse model of polycystic kidney disease. Exhibits myocardial and renal protective effects in hypertensive heart failure rats. Orally active.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type Tolvaptan
Purity >98%
Molecular Weight (g/mol) 448.94
Inhibitors Tolvaptan

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.