missing translation for 'onlineSavingsMsg'
Learn More

Riedel-de Haën Toluene CHROMASOLV™

Product Code. 15757507
Click to view available options
Quantity:
18 L
45 L
200 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Toluene CHROMASOLV™ for HPLC and organic synthesis, 99.9%.

  • For HPLC and organic synthesis.
  • High-purity, multipurpose solvents.
  • High lot-to-lot consistency.

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, phenylmethane
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

pH No data available
Melting Point -95°C
Color Colorless
CAS Max % 100.00%
Density 0.867 g/cm3 (20°C)
Boiling Point 110.6°C
Quantity 18 L
Flash Point 7.1 %(V)
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line CHROMASOLV™
Linear Formula C6H5CH3
Vapor Pressure 38 hPa (25°C)
UN Number UN1294
Viscosity No data available
Beilstein 635760
Concentration ≥99.90%
Percent Purity ≥99.90%
Grade HPLC
Chemical Name or Material Toluene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Toluene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'