Learn More
Tocris Bioscience™ Colivelin
GREENER_CHOICE
Product Code. 18025832
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
Product Code. 18025832
Brand: Tocris Bioscience™ 3945/500U
STAT3 activator; also protects against β-amyloid neurotoxicity
Colivelin is a neuroprotective peptide and activator of STAT3. Protects neurons against the neurotoxic effects of amyloid β-peptide (1-43) at a concentration of 100 fM in vitro. Suppresses neuronal death by activating STAT3 in vitro; upregulates cholinergic transmission and ameliorates memory impairment in Alzheimer's disease (AD) models. Also prevents alcohol-induced apoptosis in a fetal mouse model. Brain penetrant.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 500 μg