Learn More
Tocris Bioscience™ BQ-123
Selective ETA antagonist
Brand:  Tocris Bioscience™ 1188/500U
Product Code. 18044792
  • 267.00€ / 500µg
Explore more special offers
Description
Description
BQ-123 is a selective ETA endothelin receptor antagonist (Ki values are 1.4 and 1500 nM at ETA and ETB receptors respectively). Reduces ischemia-induced ventricular arrhythmias in a rat model.
Specifications
Specifications
0.95
Store at -20°C
500 μg