missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMI 1, >98%, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Adam 17 (TACE) and MMP inhibitor (IC50 values are 3, 4.7, 6.6, 8.4, 12, 26 and 26 nM for MMP-13, MMP-2, MMP-1, ADAM 17, MMP-9, MMP-7 and MMP-14, respectively). Suppresses TNF-α production in an acute LPS-mouse model. Reduces severity score in an in vivo model of rheumatoid arthritis. Displays selective cytotoxicity to tumor cells and cancer stem cells in vitro. Induces tumor apoptosis in a breast cancer in vivo model. Orally bioavailable.
Specifications
Specifications
Linear Formula | C17H22N2O5S2 |
Quantity | 50 mg |
Solubility Information | Soluble to 100mM in DMSO and to 20mM in ethanol |
Formula Weight | 398.49 |
Chemical Name or Material | (3S)-4-[[4-(2-Butyn-1-yloxy)phenyl]sulfonyl]-N-hydroxy-2,2-dimethyl-3-thiomorpholinecarboxamide |
Grade | >98% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'