missing translation for 'onlineSavingsMsg'
Learn More

UVP TLW&-20 Model Transilluminator

Brand:  UVP 95-0415-02

Product Code. 12883938

  • 2165.00€ / Each
Estimated Shipment: 05-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Single
365/486 nm
337 mm
200 x 200 mm
200 x 200 mm
230 V
UV/White
143 mm
UV/White Light Transilluminator
230 V 50 Hz
TLW-20
50 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.