missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SI Analytics™ TitroLine™ 7000 Basic Unit Model TL 7000-M1/20
Features the highest safety for measuring and calibrating through wireless sensor recognition for ID-electrodes. SI Analytics™ TitroLine™ 7000 Basic Unit Model TL 7000-M1/20 possesses general titration feautres but offers improved performace.
Brand: SI Analytics™ 285220150
5469.68 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
- As a rule 10 - 15 user methods are usually enough for the most requirements. But sometimes you need a little bit more capacity. The TitroLine™ 7000 can store up to 50 user methods.
- Highest safety for measuring and calibrating through wireless sensor recognition for TitroLine™ SI Analytics ID-electrodes.
- The new electrodes with definite identification are sending their specific data wireless to the titrator. Therefore the TitroLine™ 7000 uses always the correct calibration data. Erroneous measurements are excluded.
- With a pH stat application a given pH is first adjusted and then kept constant at the certain time with an acid or a base. The pH stat titration is applied to e.g.
- the determination of the enzyme activity
- the pH stat elution of soil sample at pH 4
- the monitoring of the pH value during chemical syntheses
- It is possible to detect and calculate up to 2 equivalence points during one titration. Thanks to this it is possible to determine also the Calcium and Magnesium hardness instead of only the total hardness.
Specifications
TitroLine 7000 Basic Unit | |
Polypropylene | |
10°C to 40°C | |
15.3 cm |
Brown Glass Bottle For Titration Reagent, GL 45 And S 40 Bottle Adapters, Hoses, Drip Off Tube And Titration Tip. | |
TL 7000-M1/20 | |
45 cm | |
29.6 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction