Learn More
WTW™ TISAB™ Buffer Solution
Ion strength adjustment solutions have multiple tasks: TISAB for fluoride measurements does not only adjust the pH value, by also decomplexes the fluoride-containing complex compounds. Other ion strength adjustment solutions ensure an adapted pH value as well as the addition of a suitable conducting salt to correctly capture the concentration.
Brand:  WTW™ 140100
Product Code. 11938094
  • 336.00€ / 4 L
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
Fluoride Electrode
TISAB Buffer Solution