Learn More
Lovibond™ Tintometer™ Hydrogen Peroxide LR Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
50.35€ - 105.00€
Specifications
For Use With (Application) | Photometry Tablet Reagents |
---|---|
For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
Type | Hydrogen Peroxide LR (Low Range) Tablets |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15311755
|
Lovibond™
512381BT |
250 Tablets |
105.00€
Pack of 250 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
15301755
|
Lovibond™
512380BT |
100 Tablets |
50.35€
Pack of 100 |
Estimated Shipment: 17-01-2025
Log in to see stock. |
|||||
Description
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
- Hydrogen Peroxide LR (Low Range) Tablets
- New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
- With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
- Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
- Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
- ‘BT’ is added to the end of the code to identify the new style of packaging
- Simple, time-saving and practical
- There are no safety risks if the tablets are used in line with the instructions supplied
- Safety data sheets are available for all reagents
Specifications
Photometry Tablet Reagents | |
Test kits, colorimeters and spectrophotometers | |
Hydrogen Peroxide LR (Low Range) Tablets |
Safety and Handling
- X250 HYDROGEN PEROXIDE LR TABLETS .
Signal Word
- Danger
Hazard Category
- Specific target organ toxicity Category 1
Hazard Statement
- H372-Causes damage to organs.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P314-Get medical advice/attention if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : potassium iodide
Your input is important to us. Please complete this form to provide feedback related to the content on this product.