missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(IV) Chloride Pentahydrate, 98%, Honeywell Fluka™
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Color | White to Yellow |
Quantity | 1 kg |
Linear Formula | SnCl4 · 5H2O |
UN Number | UN2440 |
Formula Weight | 350.6g/mol |
Percent Purity | 98% |
Packaging | Poly Bottle |
Appearance | Powder or Chunks |
Chemical Name or Material | Tin(IV) Chloride Pentahydrate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction