missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Timolol maleate

Product Code. 17231242
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26921-17-5
Molecular Formula C17H28N4O7S

Specifications

Quantity 250 mg
Solubility Solubility aq. soln.: soluble. Other solubilities: practically insoluble in ether, soluble in dmso, methanol, ethanol
Melting Point 202.0°C to 203.0°C
Formula Weight 432.49
InChI Key WLRMANUAADYWEA-NWASOUNVSA-N
SMILES OC(=O)\C=C/C(O)=O.CC(C)(C)NC[C@H](O)COC1=NSN=C1N1CCOCC1
IUPAC Name (2S)-1-(tert-butylamino)-3-{[4-(morpholin-4-yl)-1,2,5-thiadiazol-3-yl]oxy}propan-2-ol; (2Z)-but-2-enedioic acid
Molecular Weight (g/mol) 432.49
Chemical Name or Material Timolol maleate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'