missing translation for 'onlineSavingsMsg'
Learn More

Thrombospondin (TSP) Ab-11, Mouse Monoclonal Antibody,Lab Vision™ Epredia™

Provide accurate, reproducible results with the Epredia™ Thrombospondin (TSP) Ab-11, Mouse Monoclonal Antibody.

Brand:  Epredia MS-1066-P1ABX

 View more versions of this product

Product Code. 12658806

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.

Host Species: Mouse

Clone: D4.6 + A6.1 + MBC 200.1

Isotype: IgG1 + IgG1 + IgG1

Species Reactivity: Human. Others not known.

Immunogen: Purified human TSP

Molecular Weight: ∽450kDa (nonreduced), 170 to 180kDa (reduced)

Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting (Reduced and nonreduced)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Thrombospondin (TSP) Ab-11
Monoclonal
1mg/mL
Mouse
100 μL
Angiogenesis
Human
IgG1
Western Blot
D4.6 + A6.1 + MBC 200.1
Unconjugated
Purified human TSP
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.