Learn More
DWK Life Sciences Three Part Retaining Device, complete, for stopcock key threaded, SBW -J glass
Product Code. 10384951 Shop All DWK Life Sciences Products
Click to view available options
Closure Size:
12.5 NS
14.4 NS
18.8 NS
Unit Size:
Pack of 100
Product Code. 10384951
Brand: DWK Life Sciences 286039009
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
To secure the tap plug in the ground DURAN™ single way stopcock.
The 3-part tap safety device holds and secures the threaded tap plug in the tap sleeve.
  • 3-part, complete for SBW-J tap plug
  • Greater safety through installation of the 3-part screw locking device
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Autoclavable Not Autoclavable
Closure Size 12.5 NS
No. per Pack 100/Pk.
For Use With (Application) For securing the threaded tap plug in the ground DURAN™ one-way tap