Learn More
SCAT™ Thread Adapter, Type 60
Product Code. 14748976 Shop All SCAT Europe Products
Click to view available options
Closure Size:
GL38, GL45
Unit Size:
Each
Product Code. 14748976
Brand: SCAT™ 107339
Only null left
Add to basket
Add to basket
Safety Caps and Safety Waste Caps are available in many thread sizes. If the right thread for your container is still not available, simply use the matching adapter.
SCAT™ thread adapters are ideal for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
  • Made of polypropylene (PP) material
  • Available in a wide range of thread sizes
  • Suitable for all SCAT™ caps, funnels or canisters
Specifications
Closure Size GL38, GL45
Closure Color White
Liner Material Polypropylene
For Use With (Equipment) Politainer
Product Type Thread Adapter