Learn More
SymLine™ Adapters
Shop All SCAT Europe Products
Click to view available options
:
Adapters; Description: Thread Adapter, S90 (f) to S60 (m); Material: HDPE-EC; Type: Adapter
Description
Already exists
SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.
- Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
- All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards
Spécification
Spécification
| Closure Color | Black |
| Description | S90 (f) to S60 (m) |
| Color | Black |
| Liner Material | HDPE-EC |
| Material | HDPE-EC |
| Product Type | Thread Adapter |