missing translation for 'onlineSavingsMsg'
Learn More

Thorium solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™

Product Code. 12937433 Shop All Fisher Chemical Products
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Bottle
Unit Size:
100mL
50mL
This item is not returnable. View return policy

Product Code. 12937433

Brand: Fisher Chemical S 660 050

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula Th

Specifications

Packaging Bottle
Concentration 10 mg/L
Solvent or Matrix Nitric Acid
Quantity 50 mL
Chemical Name or Material Thorium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.