missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiocarbohydrazide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Thiocarbohydrazide is used in electron microscopy to produce electron-opaque deposits for ultra structural analysis. It is used in the synthesis of oxazine grass ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Thiocarbohydrazide is used in electron microscopy to produce electron-opaque deposits for ultra structural analysis. It is used in the synthesis of oxazine grass ketones.
Solubility
Very slightly soluble (0.5g/100g, 25°C). Insoluble in ether.
Notes
Light Sensitive. Incompatible with strong acids, Strong oxidizing agents. Avoid heat.
Thiocarbohydrazide is used in electron microscopy to produce electron-opaque deposits for ultra structural analysis. It is used in the synthesis of oxazine grass ketones.
Solubility
Very slightly soluble (0.5g/100g, 25°C). Insoluble in ether.
Notes
Light Sensitive. Incompatible with strong acids, Strong oxidizing agents. Avoid heat.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∽175°C (decomposition) |
Quantity | 25 g |
UN Number | UN2811 |
Beilstein | 506657 |
Sensitivity | Light sensitive |
Solubility Information | Very slightly soluble (0.5g/100g,25°C). Insoluble in ether. |
Formula Weight | 106.15 |
Percent Purity | 97% |
Chemical Name or Material | Thiocarbohydrazide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'