Learn More
Greiner Bio-One ThinCert™ Tissue Culture Inserts
Research tool for transport, co-culture, migration and invasion studies. Greiner Bio-One ThinCert™ Tissue Culture Inserts provides improved cell adhesion due to physical surface treatment.
Brand:  Greiner Bio-One 657631
 View more versions of this product
Product Code. 10257033
  • 270.00€ / Pack of 24
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Clear polystyrene housing with sealed PET capillary pore membrane
  • Hanging geometry provides protection to lower compartment cell layer
  • Simplified pipetting due to self-lift geometry
  • Eccentric in-well positioning allows unequalled access to lower compartment
  • Available in 6, 12 and 24 well format
TRUSTED_SUSTAINABILITY
Specifications
Specifications
452.4 mm2
2.0 to 4.15 mL
3.0 μm
6
Polystyrene
2 x 106 pores/cm2
Sterile