missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Thin Wall 0.5 mL PCR Tubes with Attached Caps
Manufactured without lubricants or chemical additives that can contaminate reagents and samples.Thin Wall 0.5mL PCR Tubes with Attached Caps are autoclavable and RNase-, DNase-, and DNA-free. Come non-sterile. Thin, uniform walls ensure consistent, precise thermal transfer. Attached flat caps form a tight seal to help prevent sample evaporation but open easily. Compatible with most thermal cyclers.
Brand: BRAND™ 781312
47.99 EUR valid until 2024-12-31
Use promo code "21977" to get your promotional price.
This item is not returnable.
View return policy
Description
Manufactured from high-quality virgin plastics for excellent, reproducible results in most PCR applications, and are manufactured without lubricants or chemical additives that can contaminate reagents and samples.- Thin, uniform walls to ensure consistent, precise thermal transfer for excellent results.
- Autoclavable and RNase-, DNase-, and DNA-free.
- Compatible with most thermal cyclers.
- Supplied with caps that form a tight seal and help prevent sample evaporation, yet are easy to open and close.
Specifications
RNase, DNase and DNA free | |
0.5 mL | |
Thermal cyclers for excellent reproducible results in most PCR applications | |
Polypropylene | |
Autoclavable | |
Single-use | |
0.5 mL |
Yellow | |
1000 Pack | |
Non-sterile | |
Flat | |
Yes | |
Tube | |
Thin Wall PCR Tube |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur