Learn More
ThermoFlex™ Recirculating Chillers
Optimized for the most demanding applications, high-performance ThermoFlex Recirculating Chillers are reliable, easy-to-use and easy-to-maintain.
Brand:  Thermo Scientific 1114010500200400
Product Code. 11955457
  • 7055.10€ / Each
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
Récupel-bijdrage uitgesloten
Description
Description
Optimized for the most demanding applications, Thermo Scientific™ ThermoFlex Recirculating Chillers are reliable, easy-to-use and easy-to-maintain. The ThermoFlex platform was developed with customer input from concept to design. This high-performance chiller platform, with cooling capacity up to 24000w/81964 BTU, is configurable to meet application demands.
Ease of Use
  • An intuitive user interface for ease of operation
  • Air and water filters that can be changed while unit is in operation
  • Innovative packaging for rapid installation
  • Quick start guide enables trouble-free system start-up
Superior Performance
  • Improved cooling capacity
  • Increased reliability
  • Ease of maintenance
Configurable Design
  • Wide range of available cooling capacities
  • Variety of available options
  • Installation flexibility
Recommended for:
  • Aerospace
  • Analytical
  • Biotech
  • Chemical
  • Industrial
  • Medical
  • Pharmaceutical
  • Research
  • University
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10°C to 40°C
1400w; Pumping option PD1; 230V 50Hz
NRTL Certified to CSA Standards; CE marked
5°C to 40°C
230 V 50 Hz
50 Hz
1.9 gal.
7.2 L
1400 W
±0.1°C
230 V