Learn More
Thermo Scientific™ ThermoFlex Chiller
Minimizes space requirements, even when compared to chillers with smaller footprints.
Brand:  Thermo Scientific™ 1214C10500200400
Product Code. 13458126
  • 9155.55€ / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Internal pressure relief valve
  • Custom temperature and pressure alarms
  • An intuitive user interface for ease of operation
  • Integrated funnel for easy filling
  • Full flow fluid filter ensures clean fluid to protect your application and maximise recirculation system life
  • Easily removable condenser grill and air filter allow for quick and simple cleaning to optimise chiller performance and maximise component life
  • Air and water filters that can be changed while unit is in operation
  • Innovative, patented packaging for rapid installation
  • Quick start guide for seamless start-up in minutes
  • Advanced design allows two sides of the unit to be blocked, allowing placement in a corner while maintaining full refrigeration performance
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2.2kW (20°C setpoint and 25°C ambient)
Suitable for a range of tap water cooling applications
230 V 50 Hz
5°C to 40°C
±0.1°C accuracy
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products