Learn More
Thermo Scientific™ Sorvall™ Legend™ Centrifuge Carts
Product Code. 10040112 Shop All Thermo Scientific Products
Click to view available options
Width (Metric):
55.1 cm
74.9 cm
Height (Metric):
29.0 cm
59.9 cm
Unit Size:
Each
Product Code. 10040112
Brand: Thermo Scientific™ 76000078
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Allow you to operate your centrifuge operation away from, or under, your primary workbench
  • For all Legend T Series centrifuges; sizes as shown in the table
  • Available in bench-high or under-cabinet models
  • With casters
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 66.5 cm
Height (Metric) 29.0 cm
Width (Metric) 55.1 cm