Learn More
Thermo Scientific™ Hypersil GOLD™ Javelin™ Guard Columns
Offer convenient, easy to use protection for analytical HPLC columns
Brand: Thermo Scientific™ 25003-014006
This item is not returnable.
View return policy
Description
- Direct-connection, fingertight design for convenience and maximum efficiency
- Patented design requires no holder, allowing quick and easy replacement
- Hypersil GOLD HPLC columns provide excellent peak shape for all analyte types
- Significant reduction in peak tailing while retaining C18 selectivity
- Excellent resolution, efficiency and sensitvity
- Confidence in the accuracy and quality of analytical data
Order Info
Javelin guards are available in other Thermo Scientific phases. Please call your Customer Service Representative for more information.
Specifications
3 μm | |
10 mm | |
4 mm | |
C18 |