Learn More
Thermo Scientific™ Hypersil GOLD™ C8 Javelin™ Guard Columns
Javelin guard columns offer convenient, easy to use protection for analytical HPLC columns
363.10€
Specifications
Particle Size 5 μm
Length (Metric) 10 mm
Stationary Phase C8
View More Specs
Products 1
Product Code Brand Diameter (Metric) Inner Particle Size Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inner Particle Size Price Quantity & Availability  
11538690
View Documents
Thermo Scientific™
25205-013006
3 mm 5 μm This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
Description
Description
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
Features of Hypersil GOLD C8 Solid Phase
  • Hypersil GOLD C8 columns provide similar selectivity to C18 columns but with reduced retention
  • Lower hydrophobicity
  • Faster separations
  • Excellent peak shape
  • High efficiency
  • Outstanding sensitivity
Order Info
Javelin guards are available in other Thermo Scientific phases. Please call your local Customer Service for more information.
Specifications
Specifications
5 μm
10 mm
C8