Learn More
Thermo Scientific™ GENESYS™ 20 Spectrophotometer Cylindrical Longpath Cell Holder
Product Code. 11861872 Shop All Thermo Scientific Products
Click to view available options
Type:
Cylindrical Cell Holder
Unit Size:
Each
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 11861872
Brand: Thermo Scientific™ undefined
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Cylindrical Longpath Cell Holder. Snap-in” holder accepts one cylindrical longpath cell up to 50mm long and 22mm in diameter.
TRUSTED_SUSTAINABILITY
Specifications
Type Cylindrical Cell Holder
For Use With (Equipment) Genesys™ 20 spectrophotometer
Pathlength Up to 50 mm