Learn More
Thermo Scientific™ Fixed Needle Autosampler Syringes, 1000uL, for Thermo Scientific™ Instruments
Tous les produits Thermo Scientific Produits
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
1 mL
missing translation for 'unitSize'
Each
Description
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Spécification
Spécification
| Needle Gauge | 23 G |
| Product Type | Gas Tight GC Autosampler Syringe, Fixed Needle |
| Volume (Metric) | 1 mL |
| Length (Metric) Needle | 65 mm |
| Needle Point Style | Side Hole |
| For Use With (Equipment) | TriPlus RSH Autosampler |