Learn More
Thermo Scientific™ Dionex™ PEEK Universal Fitting for Switchos
Product Code. 11362243 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Switchos
Unit Size:
Pack of 2
Product Code. 11362243
Brand: Thermo Scientific™ 164013
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Dionex PEEK fitting is universal for Switchos.
  • INERT
  • Long hex nut and ferrule with groove
  • I.D.: 1/16 in.
TRUSTED_SUSTAINABILITY
Specifications
Connection Type Long Hex Nut And Ferrule With Groove
For Use With (Equipment) Switchos
Material PEEK
Product Line nanoLC