Learn More
Epredia™ Carousel Cassette MicroWriter™ Accessories
Product Code. 12678136 Shop All Epredia Products
Click to view available options
For Use With (Equipment):
Microwriter Carousel Cassette
Unit Size:
Each
Product Code. 12678136
Brand: Epredia™ A78210004
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Durable systems that install in minutes
Stainless-Steel Collection Chute
  • Cassettes slide onto table in order of print
  • Stainless-Steel Collection Hopper
    • Holds 75 cassettes in order of print
    • Allows random access to all cassettes
    • System includes platform, hopper, and collection guideplate
    TRUSTED_SUSTAINABILITY
    Specifications
    For Use With (Equipment) Microwriter Carousel Cassette
    Type Collection Chute