Learn More
GA International Thermal-Transfer Cryogenic Labels for Frozen Vials and Surfaces
Product Code. 17800258
Encompass_Preferred
Click to view available options
Dimensions (LxW):
101.6 x 101.6 mm
22.2 x 22.2 mm
23.9mm x 12.7mm + 11.1 mm
25.4 x 12.7 mm
25.4 x 25.4 mm
31.8 x 22.2 mm
50.8 x 25.4 mm
50.8 x 6.4 mm
Diameter (Metric):
25.4 mm
76.2 mm
Labels per Row:
1 Label
2 Labels
Unit Size:
Pack of 1000
Pack of 2000
Pack of 300
Pack of 3000
Pack of 4000
Pack of 500
Pack of 5000
Product Code. 17800258
Brand: GA International UC52NOTC34WH
Only null left
Add to basket
Add to basket
Thermal-transfer cryogenic labels that will firmly adhere to both ambient and already frozen surfaces prior to long-term storage in liquid nitrogen.
  • Ultra-resistant adhesive ensures no flagging or detachment when stored in liquid nitrogen
  • Thermal-transfer printable for the best printout and barcode quality
  • Ideal for labeling ambient temperature as well as frozen surfaces without thawing first
  • No overlap or wrap-around required for proper adhesion
  • Wide temperature range resistance
Specifications
Adhesive Type Permanent
Color White
Finish Medium Gloss
Format Roll
Material Thermoplastic Film
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
Labels per Row 2 Labels
Quantity 4000/Pk.
Shape Rectangle with Circle
Temperature Range (English) -321°F to +250°F
Temperature Range (Metric) -196°C to +121°C
Dimensions (LxW) 23.9mm x 12.7mm + 11.1 mm
Length (English) 0.5 in.
Length (Metric) 12.7 mm
Width (English) 0.94 in.
Width (Metric) 23.9 mm
Diameter (English) 3 in.
Diameter (Metric) 76.2 mm