Learn More
Eppendorf™ Thermal printer DPU-S445
Thermal line dot printer with high printing speed and clear type face production.
Brand:  Eppendorf™ 6135011000
 View more versions of this product
Product Code. 15723275
  • 848.38€ / Each
Explore more special offers
Description
Description
  • Automatic print-out of results
  • Easy and direct connection via USB
  • Compact and lightweight
  • Easy paper orientation
  • Compatible with the BioPhotometer D30 and the BioSpectrometer product line
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BioPhotometer D30 and the BioSpectrometer product line
Thermal Dot Printer
Max printing speed: 90 mm/sec, Dots/line: 832, Resolution: 8 dots/mm
EU Plug