Learn More
Cole-Parmer™ Benchtop Heat Protector, Glass Ceramic
Keep your benchtop safe from hot labware with this Glass Ceramic Benchtop Heat Protector.
Brand:  Cole-Parmer™ 00222IZ
 View more versions of this product
Product Code. 11817242
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Plate has corner feet to prevent heat contact with benchtop.
  • Glass ceramic surface resists stains and is easy to clean.
  • Temperature range is from -392°F to +1200°F (-236°C to +649°C).
Specifications
Specifications
Benchtop Heat Protector
300 mm
1 Ea.
300 mm
300 x 300 mm